Would you like to log in again?
Middlebrook 7H10, Deep Fill Plates
Supplier: Hardy Diagnostics
W30 C34
75993-650PK 100.98 USD InStock
75993-650 10015-460
Middlebrook 7H10, Deep Fill Plates
Media for Microbiology Dehydrated Media for Microbiology
Middlebrook 7H10 Agar is recommended for use in qualitative procedures for the isolation and cultivation of Mycobacterium species.
  • Optically clear polystyrene: For distortion free viewing
  • Heavy-duty: Reduces breakage during shipping
  • Stacking rings: Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Breathable packaging: Packaged in sleeves of ten plates in a breathable bag that prevents buildup of condensation and excess moisture
Industry: Clinical
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty