Would you like to log in again?
VWR® Erlenmeyer Flask with GL 45 Screw Cap
Supplier: VWR
VWR®
1632414943250 1632414943940 1632414943500 1632414943950
75994-378CS 183.39 USD InStock
75994-378 75993-570 75994-382 75994-380
VWR® Erlenmeyer Flask with GL 45 Screw Cap
Flasks Erlenmeyer Flasks
Flask is suitable for mixing liquids.
  • Very good chemical resistance
  • Scale graduations
  • Autoclavalable
Typical uses include substance storage, media preparation, and cultivation. Made from high quality Borosilicate glass 3.3.
Tolerance: 2 mm
Ordering information: Flasks are supplied with polypropylene GL45 thread caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty