Would you like to log in again?
arium® CellPlus Ultrafilter, Sartorius Corp.
Supplier: Sartorius
arium®
H2O-CUF
75994-894EA 507.8 USD InStock
75994-894
arium® CellPlus Ultrafilter, Sartorius Corp.
Filters Liquid Process Filters
The arium® CellPlus is a point-of-use ultrafilter for efficient removal of endoxtoxins, RNase | DNase, microorganisms and particles.
  • Effective removal of RNase and DNase
  • High flow rate performance
  • Certified quality
  • Flow rate (depends on the inlet pressure and the type of system)
  • Endotoxins <0.001 EU/ml, Bacteria <1 CFU/100 ml, RNase concentration <1 pg/ml, DNase concentration <5 pg/ml
  • Effective membrane area 0.5 m² (5.4 ft.²)
Designed for arium® comfort and arium® mini ultrapure water systems, this sterile-packaged ultrafilter provides the highest safety for your critical cell culture applications. A protective bell supplied with the ultrafilter additionally prevents retrograde contamination. Moreover, the high-quality material selected for arium® CellPlus enables excellent total throughputs and optimal flow rates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty