Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Electrothermal Large Volume Heating Mantles, Cole-Parmer

Supplier: Cole Parmer
Electrothermal
36013-21
75994-950EA 4853.5 USD InStock
75994-950
Electrothermal Large Volume Heating Mantles, Cole-Parmer
Heaters Heating Mantles
Accommodate 60° funnels, pear-shaped flasks, and round-bottom flasks.

  • Bottom opening allows use with 60° funnels of various diameters
  • Accept large capacity flasks between 10 to 22 liters
  • Three separate electrical circuits let you control heating of top, middle, and bottom of flasks independently

A unique 'V-shaped' design of the heating mantle accommodates both pear-shaped and round-bottom flasks. Coiled heating element, suspended within a thermal insulating material, provides optimal heat transfer and support. Even when the heating element temperature is at 842 °F (450 °C), the outer casing remains cool to the touch.

Control heat throughout the mantle using three separate electrical circuits each controlled with its own dial switch. This enables independent temperature control of top, middle, and bottom of the flask. Two colored lights by each control dial indicate 'power on' and 'heat on'. Powder-coated aluminum housing makes it easy to clean.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty