Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Electrothermal Controlled Aluminum Heating Mantles, Cole Parmer

Supplier: Cole Parmer

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Electrothermal
03012-41 03012-31 03012-51 03012-11 03012-21
75994-970EA 530.16 USD InStock
75994-970 75994-980 75994-982 75994-978 75994-968
Electrothermal Controlled Aluminum Heating Mantles, Cole Parmer
Heaters Heating Mantles
Don't get burned, outside stays 'safe to the touch'.

  • Hold round-bottom, Erlenmeyer, and volumetric flasks
  • Maximum element temperature of 842 °F (450 °C)
  • Built-in energy regulator

Electric heating mantles incorporate a heating element suspended within a ceramic fiber insulating material for safe heat transfer. Mantles have built-in power controller with a 1 to 10 scale. Maximum temperature is 842 °F (450 °C). Two neon lights indicate 'power on' and 'heat on'.

The 50-through 1000 ml mantles are fitted with one ¹/₂" diameter support rod clamp; the 2000 through 5000 ml mantles are fitted with three ¹/₂" diameter rod clamps. Includes a 6¹/₂ ft. (2 m) power cord; 115 VAC models include U.S plug (230 VAC models do not include plug).

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty