Would you like to log in again?
COMFORT 2 UNIT BENCHTOP
Supplier: SARTORIUS CORPORATION EDI
arium* comfort II Wall mounted Unit <2ppb^ arium* comfort II Wall mounted Unit with UV, Type 1 & Type 2, Total Organic Carbon(TOC): <2ppb, with TOC Monitor, Maximum Flow Rate: 2L/min/ (5 L/H), delivers ASTM Type 1 ultrapure and Type 2 pure water, all in one system, uses proven RO & EDI technology
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COMFORT 2 UNIT BENCHTOP 75995-106 H2O-II-1-TOC-B Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty