Would you like to log in again?
DURAN® TILT, Bottle Tags, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
292435667 292435659 292435634 292435626
75995-240EA 108.04 USD InStock
75995-240 75995-242 75995-244 75995-246
DURAN® TILT, Bottle Tags, DWK Life Sciences
Labels
These bottle tags can be used on their own for colour identification or to secure the protective light shield around the bottle.
  • GL 56 bottle tags are available in four colours
  • Made of silicone
  • Rated to 200 °C
  • Autoclavable
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty