Would you like to log in again?
DURAN® Laboratory Bottles, Pressure plus+, Narrow Neck, GL 45, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
218162403
75995-262CS 542.04 USD InStock
75995-262
DURAN® Laboratory Bottles, Pressure plus+, Narrow Neck, GL 45, DWK Life Sciences
Bottles Glass Bottles
The DURAN® pressure plus+ GL 45 bottles are manufactured from DURAN® borosilicate 3.3 glass, renowned for its excellent thermal performance and high resistance to chemical attack. It offers a safer option for laboratory applications involving vacuum or pressure.
  • Vacuum filtration
  • Collection of samples under pressure
  • Storage of volatile solvents and of gas generating materials
  • Bottles are fully autoclavable and sterilizable
The pressure plus+ bottles have a guaranteed pressure resistance, and consequently offer optimal safety for both the user and contents.
Three versions of the pressure plus+ bottles are available; clear DURAN® glass with or without a protect plastic safety coating, and amber DURAN® glass for protection against ultraviolet light degradation.
Manufacturing traceability is offered through a lot specific Retrace Code printed on the bottle. Protect coated version offers protection for the user and contents in the event of breakage due to impact.
Certifications: The ambered glass meets UV light protection limits as specified in the current revisions of the USP and EP 3.2.1. The special geometry still conforms to ISO 4796-1:2016 to maintain instrument compatibility. DIN EN 1595:1997-05.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty