Would you like to log in again?
DURAN® Baffled Bottles, Wide Mouth, GLS 80®, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences
212865453 212863658 212864457
75995-266CS 59.78 USD InStock
75995-266 75995-268 75995-270
DURAN® Baffled Bottles, Wide Mouth, GLS 80®, Wheaton®, DWK Life Sciences
Bottles Roller Bottles
Baffled bottles are made up of DURAN® borosilicate 3.3 glass.
  • Resistance to chemicals
  • Easy-to-read graduated volume scale
  • Baffles are moulded on the side wall of the bottles
  • wide mouth provides easy access for cleaning
  • Compatible with magnetic stirrers and other magnetic stir bars
Bottles are applicable for aerating, dissolving, emulsifying, homogenizing, mixing and suspending of the fluids.
The baffles improve the top-to-bottom circulation and produce a greater radial mixing. The baffles also offer other advantages such as suppressing the formation of a central vortex, and reducing dead-zones where mixing does not occur.
Delivery information: Bottles are supplied complete with polypropylene pouring ring and GLS 80® cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty