Would you like to log in again?
Merit™ Sterile Foerster Sponge Forceps, Sklar
Supplier: Sklar
96-2533M
75995-394CS 1277.62 USD InStock
75995-394
Merit™ Sterile Foerster Sponge Forceps, Sklar
Forceps
Merit™ Sterile Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis.
  • Securely grips gauze and holds it in place
  • Ready-to-use
  • Office grade
The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. Merit™ Sterile instruments are ready-to-use, sterile, disposable, office-grade instruments most commonly utilized for routine and in-office minor procedures. They are sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty