Would you like to log in again?
Merit™ Sterile Halsted Mosquito Forceps, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Merit™
96-2537M 96-2539M 96-2536M 96-2538M 96-2537A
75995-396CS 473.39 USD InStock
75995-396 10194-052 75995-398 10022-490 10022-488
Merit™ Sterile Halsted Mosquito Forceps, Sklar
Forceps
Merit™ Sterile Halsted Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels.
  • Ready-to-use
  • Eliminates the need for cleaning and reprocessing
  • Significantly reduce likelihood of cross-contamination
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan
Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps, however, the Hartmann style is finer and more delicate.
Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty