Would you like to log in again?
Merit™ Sterile Adson Dressing Forceps, Physician Grade, Sklar
Supplier: Sklar
Merit™
96-2570M 96-2571M
10022-496CS 811.31 USD InStock
10022-496 75995-404
Merit™ Sterile Adson Dressing Forceps, Physician Grade, Sklar
Forceps
Sterile Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
  • Wide thumb grasp for increased precision and control
  • Ready-to-use
  • Disposable
The forceps are ready-to-use, sterile, disposable, physician-grade instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty