Would you like to log in again?
Econo™ Sterile Operating Scissors, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Econo™
96-2520A 96-2523 96-2520 96-2522 96-2597 96-2525 96-2514 96-2593 96-2592 96-2726 96-2524 96-2649 96-2521 96-2728 96-2526 96-2527 96-2649G
75995-384CS 318.34 USD InStock
75995-384 89023-956 89023-802 10185-870 89023-936 89023-804 89023-798 89023-800 75995-422 89023-794 89023-796 89023-790 75995-382 89023-792 89024-026 89023-926 89023-928
Econo™ Sterile Operating Scissors, Sklar
Scissors
Econo™ Sterile Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
  • Three tip configurations available
  • Able to cut tougher tissue
  • Ready-to-use
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty