Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Titanium Micro Dressing Forceps with Diamond Jaws, Sklar
Supplier: Sklar
98-4365T 98-4366T 98-4362T 98-4363T 98-4367T 98-4371T 98-4364T 98-4368T 98-4372T 98-4370T
75995-494EA 3109.3 USD InStock
75995-494 75995-496 75995-498 75995-500 75995-502 75995-504 75996-264 75995-506 75995-492 75995-508
Titanium Micro Dressing Forceps with Diamond Jaws, Sklar
Forceps
Sklar's® Titanium Micro Dressing Forceps are a lighter version of their stainless steel counterparts.
  • Non-magnetic
  • Lighter than stainless steel
  • Durable and corrosion resistant
  • Reusable and nonsterile
The forceps offer superior durability and corrosion resistance. Because they are non-magnetic, they are also MRI compatible.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty