Would you like to log in again?
Ball-Dissector/Probe, Sklar
Supplier: Sklar
98-7313
75995-546EA 424.14 USD InStock
75995-546
Ball-Dissector/Probe, Sklar
Dissection Needles
The Sklar® Ball Dissector #16 is used in neurosurgical procedures.
  • Reusable
  • Nonsterile
The ball-shaped end allows the dissector to probe and explore without tearing or puncturing the surrounding tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty