Sklar's® Toledo Micro Needle Holder is most commonly used to hold fine suture needles in very small sites or microsurgical procedures.
- Stainless steel
- Available with Tungsten Carbide inserts or Diamond-Coated jaws
- Reusable
- Nonsterile
These holders are available straight or curved, in various lengths, and with or without a catch. The handle has semi-cylandrical, ergonomicly designed, texturized grips for optimal comfort and utilization. Made of stainless steel. Available with Tungsten Carbide inserts or Diamond-Coated jaws. Tungsten Carbide inserts provide a secure grip on the needle, as well as an increased resistance to wear. Diamond-coated jaws provide a very secure secure grip on the needle, as well as an increased resistance to wear.