Would you like to log in again?
Titanium Castroviejo Needle Holder with Diamond Coated Jaws, Sklar
Supplier: Sklar
98-4306T 98-4314T 98-4313T 98-4311T 98-4303T 98-4315T 98-4307T 98-4302T 98-4308T 98-4300T 98-4310T 98-4301T 98-4305T 98-4312T 98-4309T 98-4304T
75995-928EA 3931.65 USD InStock
75995-928 75995-906 75995-916 75995-908 75995-918 75995-920 75995-930 75995-922 75995-932 75995-910 75995-924 75995-934 75995-912 75995-926 75995-936 75995-914
Titanium Castroviejo Needle Holder with Diamond Coated Jaws, Sklar
Forceps
Sklar's Titanium Castroviejo Needle Holders are used for micro surgery.
  • Multiple configurations available
  • Reusable and nonsterile
The instrument is available in two tip sizes (11mm x 0.8mm, 11mm x 1.2mm). The handle is round with or without catch. The product is available in 6, 7, and 8-1/4 inches and curved or straight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty