Would you like to log in again?
Samii Grasping Forceps, Sklar
Supplier: Sklar
98-9240 98-9241
75995-990EA 2329.82 USD InStock
75995-990 75995-992
Samii Grasping Forceps, Sklar
Forceps
Sklar® Samii Grasping Forceps, which may also be referred to as tumor grasping forceps, are commonly used for grasping tissue and tumors or other lesions in neurosurgical procedures.
  • Three fenestrated holes provide increased grip
  • Angled handle provides access to obscure sites
  • Reusable
  • Nonsterile
This product has a bayonet handle with three fenestrated holes to provide the user with increased grip, control, and comfort. In addition to the bayonet handle, the tips are angled left or right. This is useful for access to, and work in, extremely small, obscure sites. The tips themselves are blunt, round, and fenestrated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty