Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Toledo Micro Needle Holders, Sklar
Supplier: Sklar
98-9343 98-9312 98-9357 98-9305 98-9325 98-9331 98-9358 98-9330 98-9303 98-9350 98-9306 98-9322 98-9347 98-9353 98-9335 98-9345 98-9348 98-9351 98-9317 98-9313 98-9341 98-9310 98-9340 98-9301 98-9326 98-9337 98-9336 98-9356 98-9327 98-9324 98-9346 98-9333 98-9332 98-9342 98-9321 98-9300 98-9316 98-9315 98-9355 98-9307 98-9352 98-9311 98-9304 98-9302 98-9320 98-9314 98-9323 98-9334
75995-582EA 1918.11 USD InStock
75995-582 75995-584 75995-586 75996-008 75996-006 75995-588 75996-028 75995-580 75995-612 75996-014 75996-036 75995-578 75995-996 75995-614 75996-034 75996-012 75995-998 75996-010 75996-032 75996-030 75995-592 75995-594 75995-596 75996-018 75995-610 75995-598 75996-016 75996-038 75995-994 75995-576 75995-590 75996-040 75995-608 75996-004 75996-048 75996-026 75995-600 75996-024 75996-046 75996-002 75995-602 75996-044 75996-000 75996-022 75995-604 75996-042 75996-020 75995-606
Toledo Micro Needle Holders, Sklar
Forceps
Sklar's® Toledo Micro Needle Holder is most commonly used to hold fine suture needles in very small sites or microsurgical procedures.
  • Stainless steel
  • Available with Tungsten Carbide inserts or Diamond-Coated jaws
  • Reusable
  • Nonsterile
These holders are available straight or curved, in various lengths, and with or without a catch. The handle has semi-cylandrical, ergonomicly designed, texturized grips for optimal comfort and utilization. Made of stainless steel. Available with Tungsten Carbide inserts or Diamond-Coated jaws. Tungsten Carbide inserts provide a secure grip on the needle, as well as an increased resistance to wear. Diamond-coated jaws provide a very secure secure grip on the needle, as well as an increased resistance to wear.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty