Would you like to log in again?
Wireform Thumb Forceps, Disposable, Sterile, Sklar
Supplier: Sklar
96-8963
75996-096CS 198.12 USD InStock
75996-096
Wireform Thumb Forceps, Disposable, Sterile, Sklar
Forceps
Wireform thumb forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures.
  • Multipurpose
  • Sterile
  • Toothed
This product is straight with 1x2 teeth, and a length of 4-1/2 inches. This sterile, disposable instrument is sold in boxes of 50.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty