Would you like to log in again?
Yasargil Micro Scissors, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
98-8936 98-8924 98-2106 98-2115 98-2117 98-2111 98-2102 98-2101 98-2110 98-2112 98-2105 98-2107 98-2114 98-2116 98-2103 98-2104 98-2113 98-2108 98-2109
75996-148EA 1454.27 USD InStock
75996-148 75996-172 75996-150 75996-160 75996-170 75995-854 75996-158 75996-146 75996-168 75996-354 75996-156 75996-166 75996-352 75996-154 75996-176 75996-164 75996-174 75996-152 75996-162
Yasargil Micro Scissors, Sklar
Scissors
Sklar's® Yasargil micro scissors are a bayonet shaped instrument used for cutting and dissecting tissue.
  • Featuring two cross hands that open and close like blades
  • Can be used to spread and probe incision area
  • Reusable and nonsterile
The scissors are most common in neuro surgery, and feature two cross hands that open and close like blades. The tips can also be used to spread and probe the area of the incision. Yasargil's are availabe in a varieity of lengths, curvatures and tip configurations.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty