Would you like to log in again?
Mehdorn Applying Forceps, Sklar
Supplier: Sklar
98-4214
75996-218EA 757.09 USD InStock
75996-218
Mehdorn Applying Forceps, Sklar
Forceps
Sklar's® Medhorn forceps are used to place micro vessel clamps in position during surgical procedures to reduce blood flow.
  • Without lock
  • Reusable and nonsterile
Unlike the Biemer, the Medhorn is not available with a locking catch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty