Would you like to log in again?
Micro Surgery Scissors with Bayonet Handle, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
98-7930 98-7918 98-7920 98-7924
10185-196EA 2643.88 USD InStock
10185-196 75996-334 10185-194 10185-192
Micro Surgery Scissors with Bayonet Handle, Sklar
Scissors
These micro scissors are most commonly used in iridectomy, cataract, and capsulotomy procedures.
  • Reusable
  • Nonsterile
These double spring scissors are available with various curvatures and handle lengths. The micro blade can be seen clearly under a microscope when used as a component of surgery.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty