Would you like to log in again?
Sterile Spencer Stitch Scissors, Disposable, Sklar
Supplier: Sklar
96-2596M
75996-386CS 936.09 USD InStock
75996-386
Sterile Spencer Stitch Scissors, Disposable, Sklar
Scissors
Merit™ Sterile Spencer Stitch Scissors are primarily used for suture removal.
  • Small hook-shaped tip on one blade
  • Delicate tips for small suture removal
  • Ready-to-use
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2" and 4-1/2" and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. Merit™ Sterile instruments are ready-to-use, sterile, disposable, office-grade instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty