Would you like to log in again?
Volumetric Infusion Pump, Infusomat®, B. Braun
Supplier: B. Braun
Infusomat®
8713052U
75996-458EA 7919.05 USD InStock
75996-458
Volumetric Infusion Pump, Infusomat®, B. Braun
Pumps Syringe Pumps
The volumetric infusion pump Infusomat® Space is one of the basic building blocks of the Space system, designed for acute care adult and pediatric facilities.
  • Easy-to-learn, intuitive and user friendly
  • 1500 entry drug library available, up to 50 care units and 15 patient profiles possible
  • Alarms are audible, clearly visible and differentiated using different tones
  • Automatic rate calculation by calculating volume over time
  • Automatic bolus reduction triggered by occlusion alarm
  • Additional upstream pressure sensor detects upstream occlusions (e.g.: closed roller clamp)
  • Data lock on three safety levels (parameters and disposables lockable)
  • Clear structured display, excellent readability and LED alarm indicator with clear alarm message in display
The large volume pump allows for flexibility and easy transport within a number of care settings. Thanks to a minimal design and light weight, Space pump can be used to configure customized, tailor-made solutions as individual pumps, small therapy units, or a complex infusion system.
Equipped with an extensive drug library, easy navigation and intuitive interface, this small infusion pump packs state-of-the-art versatility that promotes efficiencies across numerous care settings. This can also be integrated into the data communications network of every advanced hospital operation.
Dose rate mode offers automatic calculation of delivery rate based on the entry of drug concentration in conjunction with the desired dose rate (e.g.: mg/kg/min). Piggyback mode offers the possibility to interrupt the current (primary) infusion temporarily in order to administer a piggyback (secondary) infusion. Device and set-based anti-free flow clamp provides maximum security against free flow when pump door is opened or infusion line removed.
Certifications: DIN EN ISO 9001 and DIN EN ISO 13485, MDD class IIb acc. to the Directive of the Council of Medical Products 93/42/EC classification defibrillation-protected; typ CF protection class in accordance with IEC/EN 60601-1, electromagnetic compatibility EMC: IEC EN 60601-1-2: 1993, IEC EN 60601-2-24: 1998.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty