Would you like to log in again?
STONE FILTER/SPARGING 10 UM HPLC MOB PHS
Supplier: Avantor
Filter/Sparging Stone 10um^ Filter/Sparging Stone, HPLC mobile phase inlet, Material: Stainless steel, Pore size: 10um, Use with 1/8thin low pressure tubing, Diameter(Outer dimension): 0.50in (12.70mm), Length: 1in (25.4mm)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STONE FILTER/SPARGING 10 UM HPLC MOB PHS 75996-482   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty