Would you like to log in again?
Methanizer (CH4izer) for Agilent GC-FID₂, Restek
Supplier: Restek
22650-EU 22650-UK 22650-CN 22650-AU 22650-US
75996-638EA 2886.05 USD InStock
75996-638 75996-636 75996-634 75996-502 75996-632
Methanizer (CH4izer) for Agilent GC-FID₂, Restek
Gas Detection Systems
Methanizer allows ppb-level GC-FID determination of CO and CO2 without expensive equipment.
  • Simple, reliable operation
  • Precise temperature control
  • Easy tube replacement
Precise temperature control ensures complete conversion of CO and CO2 to CH4
Low-level detection of carbon monoxide (CO) and carbon dioxide (CO2) is critical for many applications. Restek’s methanizer (CH4izer) allows ppb-level determination of CO and CO2 using a cost-effective GC-FID instead of more expensive instrumentation. The methanizer is factory set to 380 °C to ensure efficient and complete conversion of CO and CO2 to CH4, but the operator can easily adjust the temperature as desired with the touch of a button. This unit controls temperatures precisely and reliably to within ± 1 °C of the defined set point and the actual temperature is displayed in real time on an easy-to-read monitor. The Restek methanizer is less cumbersome than other models and is designed for easy installation and fast catalyst tube replacement so you spend more time analyzing samples and less time on maintenance.
Factory-set temperature (380°C) can be adjusted with just the touch of a button. Fast, easy catalyst tube replacement keeps maintenance time to a minimum.
Accessories information: Convenient installation kit (sold separately) includes all parts needed for quick installation into any Agilent GC.
Installation kit includes 1/8" Stainless steel metering gas valve, 10 foot x 1/8" 304 stainless steel gas supply line (rinsed and cleaned), 1/8" Swagelok tee union, 1/8" - 1/16" tube end reducer, 1/8" stainless steel Swagelok ball shut-off valve, MINICYL regulator with 1/8" fittings.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty