Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BIO-SILICO® Silicone Stoppers, Hirschmann
Supplier: Hirschmann Laborgeräte
8905952 8905924 8905932 8905922 8905928 8905942 8905912 8905915 8905917 8905919
75996-704PK 171.76 USD InStock
75996-704 75996-702 75996-712 75996-700 75996-710 75996-698 75996-696 75996-694 75996-708 75996-706
BIO-SILICO® Silicone Stoppers, Hirschmann
Stoppers Silicone Stoppers
BIO-SILICO® stoppers are manufactured in a special process which have regular pores, thus making them ideal for the preparation and sterilization of culture media
  • High degree of air permeability, similar to that of cotton plugs
  • Autoclavable
  • Resistant to chemicals
  • Water repellent and reusable
The uniform pores and good permeability to air make the stoppers ideally suitable for cultivation of aerobic microorganisms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty