Would you like to log in again?
Pails and Lids, High-Density Polyethylene, Qorpak®
Supplier: Qorpak
240131 240135 240133 PLA-03234 259746 PLA-03294 240134 240125 240172 240171 259737 240132 240130 240126
75996-734CS 22.06 USD InStock
75996-734 75990-582 89220-158 89237-502 89220-167 89220-156 89220-168 89220-165 89220-163 16218-032 89220-153 89220-170 75996-730 15712-212
Pails and Lids, High-Density Polyethylene, Qorpak®
Buckets
Durable, sealable, high-density polyethylene pails for dispensing, storage, or waste
Rounded hand grip with metal bail. The snap-on lids feature an inner rubber sealing gasket. The Reike lid has a Reike spout with a screw cap and handy pull-out spout for easy filling and emptying. EZ Tear lid has a pull tab to secure the contents during shipping and transport.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty