Would you like to log in again?
SILCOSEN® Silicone Stoppers, Hirschmann
Supplier: Hirschmann Laborgeräte
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8905528 8905522 8905755 8905542 8905566 8905576 8905515 8905720 8905532 8905612 8905765 8905828 8905538 8905840 8905622 8905740 8905512 8905510 8905517 8905552 8905730 8905524 8905835 8905519 8905617
75996-768PK 106.1 USD InStock
75996-768 75996-766 75996-688 75996-764 75996-774 75996-752 75996-772 75996-770 75996-692 75996-690 75996-758 75996-756 75996-778 75996-678 75996-754 75996-776 75996-686 75996-784 75996-762 75996-684 75996-760 75996-782 75996-682 75996-780 75996-680
SILCOSEN® Silicone Stoppers, Hirschmann
Stoppers Silicone Stoppers
SILICOSEN® stoppers are manufactured in a special process which have regular pores, thus making them ideal for the preparation and sterilization of culture media
  • Autoclavable
  • Resistant to chemicals
  • Water repellent and reusable
Lateral surface of SILICOSEN® stoppers is covered with an external skin, water repellent, low moisture evaporation, exceptional heat resistance and easy to clean.
The uniform pores and good permeability to air make the stoppers ideally suitable for cultivation of aerobic microorganisms.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty