Would you like to log in again?
VWR® Weighing Bottles with Stopper, Low Form, Cylindrical
Supplier: VWR
VWR®
264.202.12 264.202.03 264.202.04
75997-896CS 429.14 USD InStock
75997-896 75997-892 75997-894
VWR® Weighing Bottles with Stopper, Low Form, Cylindrical
Bottles Weighing Bottles
These low form weighing bottles are used for precise weighing of solids.
  • Complies ASTM - 438, TYPE-1, Boro 3.3 glass
  • These bottles are chemically resistant and mechanically strong
These weighing bottles come with a ground-plug style glass stopper which seals the contents to prevent spilling.
Manufactured from 3.3 expansion ASTM 438 Type 1 Class A standard borosilicate glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty