Would you like to log in again?
VWR® Volumetric Flasks with HDPE Stoppers, Class B
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
134.421.04 131.421.10 134.421.07 131.421.11 134.421.05 134.421.02 134.421.08 134.421.06 131.421.09
75997-966CS 185.38 USD InStock
75997-966 75997-976 75997-968 75997-978 75997-972 75997-964 75997-974 75997-980 75997-970
VWR® Volumetric Flasks with HDPE Stoppers, Class B
Flasks Volumetric Flasks
VWR® Volumetric Flasks are made from Borosilicate 3.3 glass, ASTM 438, Type 1, Class B standard.
Flasks feature a flat bottom and a pear-shaped lower area with a long neck. Etched graduation markings facilitate measurements, and the flask's mouth is equipped with a joint to accommodate an HDPE stopper. Calibration and toleration standards are defined in the standard specifications and practices: ASTM E288, ASTM E542, ASTM E694, and ISO 1042.
Flasks are calibrated 'To Contain' a precise volume at a particular temperature. They may be used for precise dilutions and preparations of standard solutions.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty