Would you like to log in again?
Puritan® Fecal Opti-Swab® Collection & Transport System, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
CB-206
75999-268CS 616.08 USD InStock
75999-268
Puritan® Fecal Opti-Swab® Collection & Transport System, Puritan Medical Products
Media for Microbiology Prepared Media for Microbiology
For the collection, transport, and preservation of enteric bacteria.
  • Ready-to-use, complete transport kit
  • Advance kit design
  • Self-centering cap
  • High-performance swab
  • Handle has a molded breakpoint
  • Shelf life - 18 months
The advance kit design is easy to use and reliabls. Self-centering cap is leakproof and facilitates easy swab removal and disposal. High-performance swab features a patented HydraFlock tip. Handle has a molded breakpoint.
Certifications: Made in USA.
Ordering information: Products are final sale.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty