Would you like to log in again?
Human IL-1β Standard TMB ELISA Development Kit, PeproTech, Inc.
Supplier: PeproTech
900-TM95 900-T95
75999-712EA 524.32 USD InStock
75999-712 75999-724
Human IL-1β Standard TMB ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Human IL-1β ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-1β in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IL-1β ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-1β in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-1β in approximately 1000 ELISA plate wells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty