Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heated Product Shelf Chambers, Labconco®

Supplier: Labconco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

780801140 780801030 780801050 780801150 780801130 780801015 780801010 780801170 780801110 780801070 780801000 780801100 780801115 780801040
76000-958EA 14168 USD InStock
76000-958 76000-936 76000-948 76000-950 76000-940 76000-952 76000-942 76000-954 76000-932 76000-944 76000-956 76000-934 76000-978 76000-946
Heated Product Shelf Chambers, Labconco®
Furniture Shelves
Heated product shelves with clear chamber for drying of bulk samples or small containers.

  • 10" diameter shelves are removable
  • Shelf temperature may be set from -20 °C (-4 °F) to 60 °C (140 °F)
  • Shelves may be placed in freezer for pre-freezing
  • Shelves are height adjustable

For use with any FreeZone Freeze Dryer except Legacy models

Each Chamber includes a 12" diameter clear acrylic cylinder, vacuum release valve and 8" diameter opening to collector on bottom of chamber. Will fit 10.12" or 14.0" diameter condensers. Short Chambers include three shelves and three temperature probes. Tall Chambers include five shelves and five temperature probes.

Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates. The 10" diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing. Each shelf is height adjustable to accommodate a broad variety of sample sizes. Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature from the Lyo-Works display.

Certifications: ETL listed. CE marked.

Ordering information: FreeZone Freeze Dryer with Lyo-Works™ OS required (not included).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty