Would you like to log in again?
BAG METAL-IN FLAT 13000 SERIES 4INX8IN
Supplier: BOTRON COMPANY INC.
Metal-In Flat Shield Bag 13000 Series 3mil 4INX8IN^ Metal-In Flat Shield Bag 13000 Series, Material: polyester dielectric and metal layer Create Faraday effect, Dissipative Outer Layer, Puncture Resistant, Metal-in, Noncorrosive, Semi Transparent, ESD Logo, Thickness: 3mil, Dimension: 4INX8IN
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BAG METAL-IN FLAT 13000 SERIES 4INX8IN 76001-098 B13048 Pack of 100 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty