Would you like to log in again?
LONG LIFE COIL CORD 12FT COIL 1/4IN SNAP
Supplier: BOTRON COMPANY INC.
Long Life Coil Cord 12Ft coil 1/4in snap^ Long Life Coil Cord, Material: Knitted 3/4in wide Nylon, 7 Strand Tinsel Wire, 5/8in Conductive Woven Thread, 1 MegOhm Built-In, Coil Cord: PVC, High quality adjustable anti static wrist strap sets, Color: Black, Dimension: 12Ft coil 1/4in snap
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LONG LIFE COIL CORD 12FT COIL 1/4IN SNAP 76001-152 B2024 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty