Would you like to log in again?
ESD Safe Coil Cords, Botron
Supplier: Botron
B2108 B2110
76001-158EA 14.99 USD InStock
76001-158 76001-156
ESD Safe Coil Cords, Botron
Grounders Grounding Wires
Coil cords are built for extended use and withstanding the labors of the work environment. It can be used in many applications from bench to personal grounding. Non-tangle and high retention coil make this a great choice for a hassle free path to ground.
  • Lighter weight for better portability
  • Reliable path to ground
  • Available in lengths of 6' and 10' long
  • Built in 1 meg Ohm ¹/₄ watt resistor at snap end
Cords are made of black PVC cord and 7 strand tinsel wire with superior strain reliefs at both ends.
Available with a spring loaded ¹/₈" (4 mm) socket connector to standard banana jack. Each cord comes with a slip on alligator clip that attaches to the banana jack for greater expansion of use.
Coil cords are ideal for personal work station grounding, wrist straps and smocks, as well as portable grounding with the included alligator clip. The non-slip banana jack provides an excellent connection to both common point and bench grounds.
Certifications: Meets S1.1 of one to five pounds breakaway force.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty