Would you like to log in again?
ESD TABLE MAT RUBBER 5FTX2.5FTX0.12IN
Supplier: BOTRON COMPANY INC.
ESD Table Mat 3 layer 5FTX2.5FTX0.12in^ ESD Table Mat, Three-Layer Design, Material: Thick Rubber, Resistant to Hot Solder, Conductive Bottom Layer, Non-Skid Backing, Reversable from Dissipative to Conductive, Chemical and Abrasion Resistant, Latex-Free, Color: green, Dimension: 5FTX2.5FTX0.12in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ESD TABLE MAT RUBBER 5FTX2.5FTX0.12IN 76001-184 B32304 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty