Would you like to log in again?
DIAMOND PLATE MAT WITH GROUND 2FTX 3FT
Supplier: BOTRON COMPANY INC.
Diamond plate mat with ground 2Ftx 5Ft 1/8in solid vinyl^ Diamond plate mat with ground, 2Ftx 5Ft 1/8in solid vinyl, Type DP, Non-slip Design, Colors; Black, Easy To Clean, Long Lasting, Non-Skid Backing, Dissipative Top Layer, ANSI EOS/ESD S4.1 RTG, reduce strain from standing on hard surfaces,
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DIAMOND PLATE MAT WITH GROUND 2FTX 3FT 76001-250 B4235 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty