Would you like to log in again?
SMOCK ESD LIGHT WEIGHT WITH SNAP X-LARGE
Supplier: BOTRON COMPANY INC.
ESD Smock Light weight with snap cuffs XL^ Smock ESD, Fabric Content: 87% polyester 13% BASF conductive carbon fibers, NEW Tuck-Strap, Snap Cuffs, Non-Static Generating Material, lightweight smock fabric provides Faraday cage effect. Color Black, Length 34-36in, Chest: 46-48in, Size: XL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SMOCK ESD LIGHT WEIGHT WITH SNAP X-LARGE 76001-410 B7325 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty