Would you like to log in again?
BAG METAL-IN FLAT SHIELD 13000 4INX2.5FT
Supplier: BOTRON COMPANY INC.
Metal-In Flat Shield Bag 13000 Series 3mil 4INX2.5Ft^ Metal-In Flat Shield Bag 13000 Series, Material: polyester dielectric and metal layer, Dissipative inner and Outer Layer, Puncture Resistant, Metal-in, Noncorrosive, Semi Transparent, ESD Logo, Thickness: 3mil, Dimension: 4INX2.5FT
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BAG METAL-IN FLAT SHIELD 13000 4INX2.5FT 76001-596 B13430 Pack of 100 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty