Would you like to log in again?
TAPE CONDUCTIVE GRID 118FTX1IN 3IN CORE
Supplier: BOTRON COMPANY INC.
Conductive grid tape 118FtX1in 3in core^ Conductive grid tape 2 mil thick copolymer with a charge decay of <.014 seconds, Acrylic Based Adhesive, Generates Less Than 50 Volts, Tensile strength: 270N/mm square, Lead-Free RoHS Compliant, Non-Corrosive, Dimension: 118FtX1in 3in core
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TAPE CONDUCTIVE GRID 118FTX1IN 3IN CORE 76001-618 B1615 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty