Would you like to log in again?
MAT V-GROOVE FLOOR GRAY 60FTX3FTX0.125IN
Supplier: BOTRON COMPANY INC.
V-Groove non-slip floor runner 60FTX3FTX0.125IN^ V-Groove non-slip floor mat, Material: 125in Thick Vinyl/Foam, Color: gray, Non-Skid Backing, Lead-Free RoHS Compliant, meets ANSI EOS/ESD S4.1 RTG, Dissipative Top Layer, excellent electricals, and durability, Dimension: 60FTX3FTX0.125IN
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MAT V-GROOVE FLOOR GRAY 60FTX3FTX0.125IN 76001-732 BV4360 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty