Would you like to log in again?
FINGER COT ESD LATEX BLACK 9IN X-LARGE
Supplier: BOTRON COMPANY INC.
Finger cot black ESD 9in XL^ Finger cot, Material: 100% latex, Dissipative, Size: XL, Thickness: 4 mil, Length: 66mm, Color: Black, Powder-free, Rolled, ESD Rated, Surface Resistivity: Avg. Is 106 - 108 ohms, Static Decay Test: 5000 to 0 volts in.01 sec, clean room safe, Length: 9in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FINGER COT ESD LATEX BLACK 9IN X-LARGE 76001-754 B6844 Gross of 5 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty