Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Color Micro (Flea) Spinbar Magnetic Stirring Bars
Supplier: VWR
VWR®
37121-6003 37121-6008 37121-6009 37121-6017 37121-6018 37121-6021 37121-6015 37121-6007 37121-6016 37121-6027 37121-6022 37121-6024 37121-6001 37121-6025 37121-6014 37121-6020 37121-6019 37121-6026 37121-6012 37121-6011 37121-6005 37121-6004
47751-498EA 7.16 USD InStock
47751-498 47751-510 47751-494 47751-504 47751-516 47751-502 76001-858 76001-848 47751-512 76001-900 76001-856 76001-846 76001-854 76001-898 76001-844 76001-852 76001-842 76001-864 47751-508 76001-850 76001-862 76001-860
VWR® Color Micro (Flea) Spinbar Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers.
  • Available in a variety of colors and sizes
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty