Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CCT036477 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-WN114-0005 BML-WN114-0025
76002-248EA 125.45 USD InStock
76002-248 76002-250
CCT036477 ≥98% (by TLC)
CCT036477
Disrupts Wnt signaling

Inhibits Wnt signaling via blocking transcription at the β-catenin level independent of CBP or p300. Inhibited growth in several cancer cell lines (HT29, HCT116, SW480, and SNU475). CCT036447 was also active in vivo where it blocked development of zebrafish and Xenopus embryos and the expression of Wnt target genes.


MW: 347.8 g/mol
CAS Number: 305372-78-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/15/71/45401571.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty