Would you like to log in again?
Arg8-Vasopressin ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-017A
76002-266EA 2320 USD InStock
76002-266
Arg8-Vasopressin ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA kit for the detection of Arg8-Vasopressin in serum, plasma and tissue culture media.
  • High sensitivity, detecting as little as 2.84 pg/mL
  • Highly specific with negligible reactivity with oxytocin
  • Simple and easy-to-use liquid color-coded reagents reduces error
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Arg8-Vasopressin in human serum, human plasma, and tissue culture media.
The Arg8-Vasopressin EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 450 nm.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, SA-HRP, TMB Substrate, Stop solution. Alternative size available: ADI-900-017A (1 x 96 wells).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty