Would you like to log in again?
Oxytocin ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
901-153A-0001
76002-268EA 3143.86 USD InStock
76002-268
Oxytocin ELISA kit, Enzo Life Sciences
Assays ELISAs
The most sensitive, specific and flexible alternative to more costly and time-consuming methods for oxytocin detection.
  • Highly specific ELISA which does not detect vasopressin, providing confidence in assay results
  • Ultra-sensitive measurement, detecting as little as 15 pg/mL oxytocin
  • Cost effective solution, faster and less costly than LC/MS
  • Easy-to-use liquid color-coded reagents and pre-coated plates minimizes errors
  • Widely published in peer reviewed literature
  • Save with bulk kit package!
For the quantitative determination of Oxytocin in culture supernatants, milk, plasma, and serum from any species. Validated sample types include cerebral spinal fluid, saliva, tissue and urine. The Oxytocin EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 405 nm. Very low reactivity with vasopressin provides confidence in assay results. Sensitive quantification of 15 pg/mL oxytocin. Ready-to-use liquid color-coded reagents reduce errors in the lab.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: ADI-900-153A (1 x 96 wells).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty