Would you like to log in again?
SCREEN-WELL® Opioid Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2814-0500 BML-2814-0100
89156-274EA 7834.61 USD InStock
89156-274 76002-330
SCREEN-WELL® Opioid Ligand Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Opioid and Sigma Ligands Plate contains 74 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Opioid and Sigma Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty